2-Amino-3-methoxybenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS36593G1 AS36593G5
ACTIAS36593G1EA 0 CHF
ACTIAS36593G1 ACTIAS36593G5
2-Amino-3-methoxybenzonitril
2-Amino-3-methoxybenzonitril
Formel: C₈H₈N₂O
MDL Number: MFCD09744022
CAS-Nummer: 148932-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR