2-Amino-5-methoxybenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS35299G1 AS35299G5
ACTIAS35299G1EA 0 CHF
ACTIAS35299G1 ACTIAS35299G5
2-Amino-5-methoxybenzonitril
2-Amino-5-methoxybenzonitril
Formel: C₈H₈N₂O
MDL Number: MFCD05149280
CAS-Nummer: 23842-82-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR