2-[2-(Dimethylamino)ethoxy]benzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS23569G1 AS23569G5
ACTIAS23569G1EA 310 CHF
ACTIAS23569G1 ACTIAS23569G5
2-[2-(Dimethylamino)ethoxy]benzonitril
2-[2-(Dimethylamino)ethoxy]benzonitril
Formel: C₁₁H₁₄N₂O
MDL Number: MFCD08690291
CAS-Nummer: 206261-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR