3-Chloro-4-(trifluoromethoxy)benzaldehyde

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Chloro-4-trifluoromethoxybenzaldehyde

AS32483G1 AS32483G5
ACTIAS32483G1EA 0 CHF
ACTIAS32483G1 ACTIAS32483G5
3-Chloro-4-(trifluoromethoxy)benzaldehyde
3-Chloro-4-(trifluoromethoxy)benzaldehyde
Formel: C8H4ClF3O2
MW: 224,5600 g/mol
MDL Number: MFCD01631561
CAS-Nummer: 83279-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR