4-Brom-1-iod-2-nitrobenzol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Bromo-2-iodonitrobenzene

AS35288G1 AS35288G10 AS35288G5
ACTIAS35288G1EA 0 CHF
ACTIAS35288G1 ACTIAS35288G10 ACTIAS35288G5
4-Brom-1-iod-2-nitrobenzol
4-Brom-1-iod-2-nitrobenzol
Formel: C₆H₃BrINO₂
MW: 327.9020 g/mol
MDL Number: MFCD11505492
CAS-Nummer: 112671-42-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR