3-(Methylcarbamoyl)phenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-(N-Methylaminocarbonyl)phenylboronic acid, 3-(Methylcarbamoyl)benzeneboronic acid, (3-(Methylcarbamoyl)phenyl)boronic acid

AS35445G1 AS35445G5
ACTIAS35445G1EA 0 CHF
ACTIAS35445G1 ACTIAS35445G5
3-(Methylcarbamoyl)phenylboronsäure
3-(Methylcarbamoyl)phenylboronsäure
Formel: C₈H₁₀BNO₃
MW: 178,98 g/mol
MDL Number: MFCD04038918
CAS-Nummer: 832695-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR