(3-Chlor-2-nitrophenyl)essigsäure

Lieferant: ACTIVATE SCIENTIFIC

AS21776G1 AS21776G5
ACTIAS21776G1EA 0 CHF
ACTIAS21776G1 ACTIAS21776G5
(3-Chlor-2-nitrophenyl)essigsäure
(3-Chlor-2-nitrophenyl)essigsäure
Formel: C₈H₆ClNO₄
MDL Number: MFCD03424331
CAS-Nummer: 23066-21-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR