(3-Brom-2-chlorphenyl)essigsäure

Lieferant: ACTIVATE SCIENTIFIC

AS34373G1 AS34373G5
ACTIAS34373G1EA 0 CHF
ACTIAS34373G1 ACTIAS34373G5
(3-Brom-2-chlorphenyl)essigsäure
(3-Brom-2-chlorphenyl)essigsäure
Formel: C₈H₆BrClO₂
MDL Number: MFCD09864663
CAS-Nummer: 786652-63-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR