(2-Brom-4-chlorphenyl)essigsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Bromo-4-chlorophenylacetic acid

AS35546G1 AS35546G5
ACTIAS35546G1EA 0 CHF
ACTIAS35546G1 ACTIAS35546G5
(2-Brom-4-chlorphenyl)essigsäure
(2-Brom-4-chlorphenyl)essigsäure
Formel: C₈H₆BrClO₂
MW: 249.4900 g/mol
MDL Number: MFCD09032951
CAS-Nummer: 52864-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR