5-(4-Nitrophenyl)oxazol

Lieferant: ACTIVATE SCIENTIFIC

AS20801G1 AS20801G5
ACTIAS20801G1EA 68 CHF
ACTIAS20801G1 ACTIAS20801G5
5-(4-Nitrophenyl)oxazol
5-(4-Nitrophenyl)oxazole
Formel: C₉H₆N₂O₃
MW: 190,1600 g/mol
MDL Number: MFCD00085148
CAS-Nummer: 1014-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR