5-(3-Nitrophenyl)oxazol
Lieferant: ACTIVATE SCIENTIFIC
AS21830G1
AS21830G5
ACTIAS21830G1EA
0
CHF
ACTIAS21830G1
ACTIAS21830G5
5-(3-Nitrophenyl)oxazol
5-(3-Nitrophenyl)oxazole
Formel:
C₉H₆N₂O₃ MW: 190,155 g/mol |
MDL Number:
MFCD00085147 CAS-Nummer: 89808-77-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...