4-(4-Nitrophenyl)oxazol

Lieferant: ACTIVATE SCIENTIFIC

AS36457G1 AS36457G5
ACTIAS36457G1EA 0 CHF
ACTIAS36457G1 ACTIAS36457G5
4-(4-Nitrophenyl)oxazol
4-(4-Nitrophenyl)oxazole
Formel: C₉H₆N₂O₃
MDL Number: MFCD09032979
CAS-Nummer: 13382-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR