5-(3-Bromphenyl)oxazol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-(3-Bromophenyl)-1,3-oxazole

AS20852G1 AS20852G25 AS20852G5
ACTIAS20852G1EA 0 CHF
ACTIAS20852G1 ACTIAS20852G25 ACTIAS20852G5
5-(3-Bromphenyl)oxazol
5-(3-Bromophenyl)oxazole
Formel: C₉H₆BrNO
MDL Number: MFCD05668970
CAS-Nummer: 243455-57-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR