5-(2-Bromphenyl)oxazol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-(2-Bromophenyl)-1,3-oxazole

AS23737G1 AS23737G25 AS23737G5
ACTIAS23737G1EA 39 CHF
ACTIAS23737G1 ACTIAS23737G25 ACTIAS23737G5
5-(2-Bromphenyl)oxazol
5-(2-Bromophenyl)oxazole
Formel: C₉H₆BrNO
MW: 224,0540 g/mol
MDL Number: MFCD05668985
CAS-Nummer: 328270-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR