2-(2-Bromphenyl)oxazol

Lieferant: ACTIVATE SCIENTIFIC

AS30523G1 AS30523G5
ACTIAS30523G1EA 0 CHF
ACTIAS30523G1 ACTIAS30523G5
2-(2-Bromphenyl)oxazol
2-(2-Bromophenyl)oxazole
Formel: C₉H₆BrNO
MDL Number: MFCD08234434
CAS-Nummer: 92346-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR