Ethyl-2-methylthiazol-5-carboxylat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-(Ethoxycarbonyl)-2-methyl-1,3-thiazole, Ethyl 2-methylthiazole-5-carboxylate, Ethyl 2-methyl-1,3-thiazole-5-carboxylate

AS50353G1 AS50353G5
ACTIAS50353G1EA 0 CHF
ACTIAS50353G1 ACTIAS50353G5
Ethyl-2-methylthiazol-5-carboxylat
Ethyl-2-methylthiazol-5-carboxylat
Formel: C₇H₉NO₂S
MW: 171,2200 g/mol
MDL Number: MFCD03840442
CAS-Nummer: 79836-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR