Ethyl-2-methylthiazol-4-carboxylat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Ethyl 2-methylthiazole-4-carboxylate

AS50040G1 AS50040G5
ACTIAS50040G1EA 0 CHF
ACTIAS50040G1 ACTIAS50040G5
Ethyl-2-methylthiazol-4-carboxylat
Ethyl-2-methylthiazol-4-carboxylat
Formel: C₇H₉NO₂S
MW: 171,22 g/mol
MDL Number: MFCD00156148
CAS-Nummer: 6436-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR