Methylthiazol-5-carboxylat

Lieferant: ACTIVATE SCIENTIFIC

AS50095G1 AS50095G5
ACTIAS50095G1EA 0 CHF
ACTIAS50095G1 ACTIAS50095G5
Methylthiazol-5-carboxylat
Methylthiazol-5-carboxylat
Formel: C₅H₅NO₂S
MW: 143,1600 g/mol
MDL Number: MFCD03788563
CAS-Nummer: 14527-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR