Ethylthiazol-5-carboxylat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Ethyl 5-thiazolecarboxylate

AS31818G1 AS31818G10 AS31818G5
ACTIAS31818G1EA 0 CHF
ACTIAS31818G1 ACTIAS31818G10 ACTIAS31818G5
Ethylthiazol-5-carboxylat
Ethylthiazol-5-carboxylat
Formel: C₆H₇NO₂S
MW: 157,1900 g/mol
MDL Number: MFCD06205069
CAS-Nummer: 32955-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR