Ethylthiazol-2-carboxylat

Lieferant: ACTIVATE SCIENTIFIC

AS23357G1 AS23357G5
ACTIAS23357G1EA 75 CHF
ACTIAS23357G1 ACTIAS23357G5
Ethylthiazol-2-carboxylat
Ethylthiazol-2-carboxylat
Formel: C₆H₇NO₂S
MW: 157,1900 g/mol
MDL Number: MFCD06204689
CAS-Nummer: 14527-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR