5-Oxazolcarbonsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Oxazole-5-carboxylic acid

AS1818G1 AS1818G5
ACTIAS1818G1EA 0 CHF
ACTIAS1818G1 ACTIAS1818G5
5-Oxazolcarbonsäure
5-Oxazolecarboxylic acid
Formel: C₄H₃NO₃
MDL Number: MFCD04114931
CAS-Nummer: 118994-90-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR