Ethyl-2-amino-4-oxazolcarboxylat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Ethyl 2-aminooxazole-4-carboxylate

AS50052G1 AS50052G5
ACTIAS50052G1EA 0 CHF
ACTIAS50052G1 ACTIAS50052G5
Ethyl-2-amino-4-oxazolcarboxylat
Ethyl-2-amino-4-oxazolcarboxylat
Formel: C₆H₈N₂O₃
MW: 156.14 g/mol
MDL Number: MFCD06659907
CAS-Nummer: 177760-52-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR