Ethyl-4-oxazolcarboxylat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Ethyl oxazole-4-carboxylate

AS50105G1 AS50105G5
ACTIAS50105G1EA 0 CHF
ACTIAS50105G1 ACTIAS50105G5
Ethyl-4-oxazolcarboxylat
Ethyl-4-oxazolcarboxylat
Formel: C₆H₇NO₃
MW: 141,1300 g/mol
MDL Number: MFCD04114940
CAS-Nummer: 23012-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR