Ethyl-2-chlor-5-oxazolcarboxylat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Ethyl 2-chlorooxazole-5-carboxylate

AS1919G1 AS1919G5
ACTIAS1919G1EA 0 CHF
ACTIAS1919G1 ACTIAS1919G5
Ethyl-2-chlor-5-oxazolcarboxylat
Ethyl-2-chlor-5-oxazolcarboxylat
Formel: C₆H₆ClNO₃
MW: 175,5697 g/mol
MDL Number: MFCD09475866
CAS-Nummer: 862599-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR