Ethyl-2-chlor-4-oxazolcarboxylat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Ethyl 2-chlorooxazole-4-carboxylate

AS50045G1 AS50045G5
ACTIAS50045G1EA 0 CHF
ACTIAS50045G1 ACTIAS50045G5
Ethyl-2-chlor-4-oxazolcarboxylat
Ethyl-2-chlor-4-oxazolcarboxylat
Formel: C₆H₆ClNO₃
MW: 175.57 g/mol
MDL Number: MFCD06660120
CAS-Nummer: 460081-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR