5-Amino-3-(3-fluorphenyl)isoxazol

Lieferant: ACTIVATE SCIENTIFIC

AS23802G1 AS23802G5
ACTIAS23802G1EA 245 CHF
ACTIAS23802G1 ACTIAS23802G5
5-Amino-3-(3-fluorphenyl)isoxazol
5-Amino-3-(3-fluorophenyl)isoxazole
Formel: C₉H₇FN₂O
MDL Number: MFCD07787075
CAS-Nummer: 119162-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR