3-(4-Methoxyphenyl)-5-isoxazolcarbonsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-(4-Methoxyphenyl)isoxazole-5-carboxylic acid

AS8376G1 AS8376G5
ACTIAS8376G1EA 0 CHF
ACTIAS8376G1 ACTIAS8376G5
3-(4-Methoxyphenyl)-5-isoxazolcarbonsäure
3-(4-Methoxyphenyl)-5-isoxazolecarboxylic acid
Formel: C₁₁H₉NO₄
MW: 219,1900 g/mol
MDL Number: MFCD04124288
CAS-Nummer: 618383-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR