5-Amino-3-(3-chlorphenyl)isoxazol

Lieferant: ACTIVATE SCIENTIFIC

AS23797G25 AS23797G5
ACTIAS23797G25EA 0 CHF
ACTIAS23797G25 ACTIAS23797G5
5-Amino-3-(3-chlorphenyl)isoxazol
5-Amino-3-(3-chlorophenyl)isoxazole
Formel: C₉H₇ClN₂O
MDL Number: MFCD07383023
CAS-Nummer: 86685-95-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR