5-Amino-3-(3,5-dichlorphenyl)isoxazol

Lieferant: ACTIVATE SCIENTIFIC

AS23803G1 AS23803G5
ACTIAS23803G1EA 0 CHF
ACTIAS23803G1 ACTIAS23803G5
5-Amino-3-(3,5-dichlorphenyl)isoxazol
5-Amino-3-(3,5-dichlorophenyl)isoxazole
Formel: C₉H₆Cl₂N₂O
MDL Number: MFCD11156303
CAS-Nummer: 1020997-14-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR