5-Amino-3-(3,4-dichlorphenyl)isoxazol
Lieferant: ACTIVATE SCIENTIFIC
AS23810G1
AS23810G5
ACTIAS23810G1EA
0
CHF
ACTIAS23810G1
ACTIAS23810G5
5-Amino-3-(3,4-dichlorphenyl)isoxazol
5-Amino-3-(3,4-dichlorophenyl)isoxazole
Formel:
C₉H₆Cl₂N₂O |
MDL Number:
MFCD07383039 CAS-Nummer: 501902-20-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...