5-Amino-3-(4-bromphenyl)isoxazol

Lieferant: ACTIVATE SCIENTIFIC

AS23796G1 AS23796G5
ACTIAS23796G1EA 0 CHF
ACTIAS23796G1 ACTIAS23796G5
5-Amino-3-(4-bromphenyl)isoxazol
5-Amino-3-(4-bromophenyl)isoxazole
Formel: C₉H₇BrN₂O
MDL Number: MFCD06199354
CAS-Nummer: 119162-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR