3-Brom-5-(4-fluorphenyl)isoxazol

Lieferant: ACTIVATE SCIENTIFIC

AS51154G1 AS51154G5
ACTIAS51154G1EA 0 CHF
ACTIAS51154G1 ACTIAS51154G5
3-Brom-5-(4-fluorphenyl)isoxazol
3-Bromo-5-(4-fluorophenyl)isoxazole
Formel: C₉H₅BrFNO
MDL Number: MFCD06660142
CAS-Nummer: 903130-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR