3-(Hydroxymethyl)-5-methylisoxazol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Hydroxymethyl-5-methylisoxazole, 5-Methyl-3-isoxazolemethanol

AS35666G1 AS35666G5
ACTIAS35666G1EA 0 CHF
ACTIAS35666G1 ACTIAS35666G5
3-(Hydroxymethyl)-5-methylisoxazol
3-(Hydroxymethyl)-5-methylisoxazole
Formel: C₅H₇NO₂
MW: 113,11 g/mol
MDL Number: MFCD00085129
CAS-Nummer: 35166-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR