3,5-Dimethyl-4-isoxazolboronsäurepinakolester

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)isoxazole, 3,5-dimethyl-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2-oxazole

AS12573G25 AS12573G5
ACTIAS12573G25EA 108 CHF
ACTIAS12573G25 ACTIAS12573G5
3,5-Dimethyl-4-isoxazolboronsäurepinakolester
3,5-Dimethyl-4-isoxazolboronsäurepinakolester
Formel: C₁₁H₁₈BNO₃
MW: 223,08 g/mol
MDL Number: MFCD05863910
CAS-Nummer: 832114-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR