3-Brom-5-(hydroxymethyl)isoxazol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (3-Bromoisoxazol-5-yl)methanol

AS50108G1 AS50108G5
ACTIAS50108G1EA 0 CHF
ACTIAS50108G1 ACTIAS50108G5
3-Brom-5-(hydroxymethyl)isoxazol
3-Bromo-5-(hydroxymethyl)isoxazole
Formel: C₄H₄BrNO₂
MW: 177,98 g/mol
MDL Number: MFCD04035586
CAS-Nummer: 25742-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR