3-Brom-5-(chlormethyl)isoxazol

Lieferant: ACTIVATE SCIENTIFIC

AS50064G1 AS50064G5
ACTIAS50064G1EA 0 CHF
ACTIAS50064G1 ACTIAS50064G5
3-Brom-5-(chlormethyl)isoxazol
3-Bromo-5-(chloromethyl)isoxazole
Formel: C₄H₃BrClNO
MDL Number: MFCD06660137
CAS-Nummer: 124498-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR