4-Nitroisoxazol

Lieferant: ACTIVATE SCIENTIFIC

AS30902G1 AS30902G5
ACTIAS30902G1EA 0 CHF
ACTIAS30902G1 ACTIAS30902G5
4-Nitroisoxazol
4-Nitroisoxazole
Formel: C₃H₂N₂O₃
MDL Number: MFCD06638027
CAS-Nummer: 1121-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR