6-Methoxy-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 6-Methoxyindazole

AS30626G1 AS30626G5
ACTIAS30626G1EA 0 CHF
ACTIAS30626G1 ACTIAS30626G5
6-Methoxy-1H-indazol
6-Methoxy-1H-indazole
Formel: C₈H₈N₂O
MW: 148,1619 g/mol
MDL Number: MFCD09261132
CAS-Nummer: 3522-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR