5-Methoxy-1H-indazol
Lieferant: ACTIVATE SCIENTIFIC
AS12288G1
AS12288G5
ACTIAS12288G1EA
0
CHF
ACTIAS12288G1
ACTIAS12288G5
5-Methoxy-1H-indazol
5-Methoxy-1H-indazole
Formel:
C₈H₈N₂O MW: 148.1600 g/mol |
MDL Number:
MFCD07781657 CAS-Nummer: 94444-96-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...