3-Iod-6-methoxy-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

AS30576G1 AS30576G5
ACTIAS30576G1EA 0 CHF
ACTIAS30576G1 ACTIAS30576G5
3-Iod-6-methoxy-1H-indazol
3-Iodo-6-methoxy-1H-indazole
Formel: C₈H₇IN₂O
MW: 274.0585 g/mol
MDL Number: MFCD09266218
CAS-Nummer: 936138-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR