3-Iod-6-nitro-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

AS51815G1 AS51815G5
ACTIAS51815G1EA 0 CHF
ACTIAS51815G1 ACTIAS51815G5
3-Iod-6-nitro-1H-indazol
3-Iodo-6-nitro-1H-indazole
Formel: C₇H₄IN₃O₂
MDL Number: MFCD07781652
CAS-Nummer: 70315-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR