3-Amino-4-iod-1H-indazol
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
4-Iodo-1H-indazol-3-amine
AS14049G1
AS14049G5
ACTIAS14049G1EA
0
CHF
ACTIAS14049G1
ACTIAS14049G5
3-Amino-4-iod-1H-indazol
3-Amino-4-iodo-1H-indazole
Formel:
C₇H₆IN₃ MW: 259.0471 g/mol |
MDL Number:
MFCD03426697 CAS-Nummer: 599191-73-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...