Methyl-3-iod-1H-indazol-5-carboxylat
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
Methyl 3-iodo-1H-indazole-5-carboxylate
AS1443G1
AS1443G5
ACTIAS1443G1EA
0
CHF
ACTIAS1443G1
ACTIAS1443G5
Methyl-3-iod-1H-indazol-5-carboxylat
Methyl-3-iod-1H-indazol-5-carboxylat
Formel:
C₉H₇IN₂O₂ MW: 302,0700 g/mol |
MDL Number:
MFCD06739144 CAS-Nummer: 885271-25-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...