Methyl-3-iod-1H-indazol-4-carboxylat

Lieferant: ACTIVATE SCIENTIFIC

AS1640G1 AS1640G5
ACTIAS1640G1EA 0 CHF
ACTIAS1640G1 ACTIAS1640G5
Methyl-3-iod-1H-indazol-4-carboxylat
Methyl-3-iod-1H-indazol-4-carboxylat
Formel: C₉H₇IN₂O₂
MDL Number: MFCD07781588
CAS-Nummer: 885521-54-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR