6-Iod-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 6-Iodoindazole

AS12291G1 AS12291G25 AS12291G5
ACTIAS12291G1EA 0 CHF
ACTIAS12291G1 ACTIAS12291G25 ACTIAS12291G5
6-Iod-1H-indazol
6-Iodo-1H-indazole
Formel: C₇H₅IN₂
MW: 244.0300 g/mol
MDL Number: MFCD04114695
CAS-Nummer: 261953-36-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR