5-Iod-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

AS31622G1 AS31622G5
ACTIAS31622G1EA 0 CHF
ACTIAS31622G1 ACTIAS31622G5
5-Iod-1H-indazol
5-Iodo-1H-indazole
Formel: C₇H₅IN₂
MDL Number: MFCD07781642
CAS-Nummer: 55919-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR