4-Iod-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

AS12258G1 AS12258G5
ACTIAS12258G1EA 0 CHF
ACTIAS12258G1 ACTIAS12258G5
4-Iod-1H-indazol
4-Iodo-1H-indazole
Formel: C₇H₅IN₂
MDL Number: MFCD07781607
CAS-Nummer: 885522-11-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR