3-Iod-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Iodoindazole

AS30974G1 AS30974G5
ACTIAS30974G1EA 0 CHF
ACTIAS30974G1 ACTIAS30974G5
3-Iod-1H-indazol
3-Iodo-1H-indazole
Formel: C₇H₅IN₂
MW: 244.0325 g/mol
MDL Number: MFCD04972469
CAS-Nummer: 66607-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR