3-Amino-4,6-difluor-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

AS22567G1 AS22567G5
ACTIAS22567G1EA 0 CHF
ACTIAS22567G1 ACTIAS22567G5
3-Amino-4,6-difluor-1H-indazol
3-Amino-4,6-difluoro-1H-indazole
Formel: C₇H₅F₂N₃
MDL Number: MFCD13191634
CAS-Nummer: 1197193-24-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR