5-Fluor-3-iod-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

AS1896G1 AS1896G5
ACTIAS1896G1EA 0 CHF
ACTIAS1896G1 ACTIAS1896G5
5-Fluor-3-iod-1H-indazol
5-Fluoro-3-iodo-1H-indazole
Formel: C₇H₄FIN₂
MW: 262.02 g/mol
MDL Number: MFCD06739145
CAS-Nummer: 858629-06-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR